3-Hydroxy-4-iodbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

664057-1G
SIAL664057-1GEA 73.3 EUR
SIAL664057-1G
3-Hydroxy-4-iodbenzoesäure, Sigma-Aldrich®
3-Hydroxy-4-iodbenzoesäure
Formel: C₇H₅IO₃
Molecular Weight: 264,02 g/mol
MDL: MFCD02068386
CAS-Nummer: 58123-77-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR