(4-Methylphenyl)methansulfonsäurechlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: p-Tolylmethanesulphonyl chloride, α-p-Xylenesulphonyl chloride, 4-Methylbenzylsulphonyl chloride

M-Clarity™ Qualitätsstufe = MQ100

664766-1G
SIAL664766-1GEA 93.8 EUR
SIAL664766-1G
(4-Methylphenyl)methansulfonsäurechlorid, Sigma-Aldrich®
(4-Methylphenyl)methanesulphonyl chloride
Formel: C₈H₉ClO₂S
Molecular Weight: 204.67 g/mol
MDL: MFCD01631931
CAS-Nummer: 51419-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR