(4-Chlorphenyl)methansulfonsäurechlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Chloro-α-toluenesulphonyl chloride

M-Clarity™ Qualitätsstufe = MQ100

664774-1G
SIAL664774-1GEA 41.2 EUR
SIAL664774-1G
(4-Chlorphenyl)methansulfonsäurechlorid, Sigma-Aldrich®
(4-Chlorophenyl)methanesulphonyl chloride
Formel: C₇H₆Cl₂O₂S
Molecular Weight: 225,10 g/mol
MDL: MFCD01631927
CAS-Nummer: 6966-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR