4-Bromanthranilsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-Amino-4-bromobenzoic acid

M-Clarity™ Qualitätsstufe = MQ100

664863-5G
SIAL664863-5GEA 163 EUR
SIAL664863-5G
4-Bromanthranilsäure, Sigma-Aldrich®
4-Bromanthranilsäure
Formel: C₇H₆BrNO₂
Molecular Weight: 216.03 g/mol
MDL: MFCD03618454
CAS-Nummer: 20776-50-5
EINECS: 244-025-7
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR