Methyl-5-iod-2-methoxybenzoat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-Iod-2-methoxybenzoesäuremethylester
M-Clarity™ Qualitätsstufe = MQ100
665304-10G
SIAL665304-10GEA
376
EUR
SIAL665304-10G
Methyl-5-iod-2-methoxybenzoat, Sigma-Aldrich®
Methyl-5-iod-2-methoxybenzoat
Formel:
C₉H₉IO₃ Molecular Weight: 292.07 g/mol |
MDL:
MFCD02683862 CAS-Nummer: 40757-09-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...