2-Brom-5-iodbenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

665339-10G
SIAL665339-10GEA 258 EUR
SIAL665339-10G
2-Brom-5-iodbenzoesäure, Sigma-Aldrich®
2-Brom-5-iodbenzoesäure
Formel: C₇H₄BrIO₂
Molecular Weight: 326,92 g/mol
MDL: MFCD00079716
CAS-Nummer: 25252-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR