4-(Trifluormethyl)piperidin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

665509-1G
SIAL665509-1GEA 108 EUR
SIAL665509-1G
4-(Trifluormethyl)piperidin Hydrochlorid, Sigma-Aldrich®
4-(Trifluormethyl)piperidin Hydrochlorid
Formel: C₆H₁₁ClF₃N
Molecular Weight: 189.61 g/mol
MDL: MFCD04971993
CAS-Nummer: 155849-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR