4-(Methylamino)benzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

665843-5G
SIAL665843-5GEA 174 EUR
SIAL665843-5G
4-(Methylamino)benzonitril, Sigma-Aldrich®
4-(Methylamino)benzonitril
Formel: C₈H₈N₂
Molecular Weight: 132,16 g/mol
MDL: MFCD00016381
CAS-Nummer: 4714-62-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR