Methyl-2-benzo[b]thiophencarboxylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

666432-10G
SIAL666432-10GEA 118 EUR
SIAL666432-10G
Methyl-2-benzo[b]thiophencarboxylat, Sigma-Aldrich®
Methyl-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₈O₂S
Molecular Weight: 192.24 g/mol
MDL: MFCD00067791
CAS-Nummer: 22913-24-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR