5-Indolboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (1H-indol-5-yl)boronic acid, (1H-Indol-5-yl)boronic acid, 5-Indolylboronic acid

M-Clarity™ Qualitätsstufe = MQ100

666467-1G
SIAL666467-1GEA 49.2 EUR
SIAL666467-1G
5-Indolboronsäure, Sigma-Aldrich®
5-Indoleboronic acid
Formel: C₈H₈BNO₂
MDL: MFCD01319013
CAS-Nummer: 144104-59-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR