2,4-Dimethoxy-5-pyrimidinboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4-Dimethoxypyrimidine-5-boronic acid, (2,4-Dimethoxypyrimidin-5-yl)boronic acid, 2,4-Dimethoxy-5-pyrimidylboronic acid

M-Clarity™ Qualitätsstufe = MQ100

666491-1G
SIAL666491-1GEA 45 EUR
SIAL666491-1G
2,4-Dimethoxy-5-pyrimidinboronsäure, Sigma-Aldrich®
2,4-Dimethoxy-5-pyrimidineboronic acid
Formel: C₆H₉BN₂O₄
Molecular Weight: 183,9577 g/mol
MDL: MFCD01318985
CAS-Nummer: 89641-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR