5-Chlorbenzoxazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

666602-5G
SIAL666602-5GEA 83.7 EUR
SIAL666602-5G
5-Chlorbenzoxazol, Sigma-Aldrich®
5-Chlorobenzoxazole
Formel: C₇H₄ClNO
Molecular Weight: 153,57 g/mol
MDL: MFCD00216939
CAS-Nummer: 17200-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR