4-Methoxy-2-(trifluormethyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methoxy-2-(trifluoromethyl)benzeneboronic acid, (4-Methoxy-2-(trifluoromethyl)phenyl)boronic acid

M-Clarity™ Qualitätsstufe = MQ100

666912-1G
SIAL666912-1GEA 77 EUR
SIAL666912-1G
4-Methoxy-2-(trifluormethyl)phenylboronsäure, Sigma-Aldrich®
4-Methoxy-2-(trifluormethyl)phenylboronsäure
Formel: C₈H₈BF₃O₃
Molecular Weight: 219.95 g/mol
MDL: MFCD03095346
CAS-Nummer: 313546-16-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR