3-Amino-5-brompyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 5-Brom-3-pyridylamin, 5-Brompyridin-3-amin

M-Clarity™ Qualitätsstufe = MQ100

667129-10G
SIAL667129-10GEA 43.2 EUR
SIAL667129-10G
3-Amino-5-brompyridin, Sigma-Aldrich®
3-Amino-5-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173.01 g/mol
MDL: MFCD01646060
CAS-Nummer: 13535-01-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR