2,3,4-Trichlorbenzolsulfonylchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,3,4-Trichlorbenzol-1-sulfonylchlorid, 2,3,4-Trichlorbenzolsulfonsäurechlorid, 2,3,4-Trichlorbenzolsulfochlorid
M-Clarity™ Qualitätsstufe = MQ100
667439-5G
SIAL667439-5GEA
102
EUR
SIAL667439-5G
2,3,4-Trichlorbenzolsulfonylchlorid, Sigma-Aldrich®
2,3,4-Trichlorbenzolsulfonylchlorid
Formel:
C₆H₂Cl₄O₂S Molecular Weight: 279,96 g/mol Schmelzpunkt: 65…66 °C |
MDL:
MFCD00024871 CAS-Nummer: 34732-09-7 EINECS: 252-174-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...