3-Methyl-2-furancarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Methylfuran-2-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

667994-1G
SIAL667994-1GEA 57.9 EUR
SIAL667994-1G
3-Methyl-2-furancarbonsäure, Sigma-Aldrich®
3-Methyl-2-furoic acid
Formel: C₆H₆O₃
Molecular Weight: 126.11 g/mol
MDL: MFCD00014111
CAS-Nummer: 4412-96-8
EINECS: 224-571-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR