4-(3-Pyridyl)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Pyridin-3-yl)benzaldehyde

M-Clarity™ Qualitätsstufe = MQ400

668303-1G
SIAL668303-1GEA 79.6 EUR
SIAL668303-1G
4-(3-Pyridyl)benzaldehyd, Sigma-Aldrich®
4-(3-Pyridyl)benzaldehyd
Formel: C₁₂H₉NO
Molecular Weight: 183,2060 g/mol
MDL: MFCD01631910
CAS-Nummer: 127406-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR