9-Undecin-1-ol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

668338-1G
SIAL668338-1GEA 114 EUR
SIAL668338-1G
9-Undecin-1-ol, Sigma-Aldrich®
9-Undecin-1-ol
Formel: C₁₁H₂₀O
MDL: MFCD04116376
CAS-Nummer: 177961-61-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR