Tetrakis(dimethylamido)titan ≥99,999%, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Tetrakis(dimethylamido)titan(IV), Tetrakis(dimethylamino)titan(IV), TDMAT, Tetrakis(dimethylamino)titan

M-Clarity™ Qualitätsstufe = MQ100

669008-25G
SIAL669008-25GEA 1090 EUR
SIAL669008-25G
Tetrakis(dimethylamido)titan ≥99,999%, Sigma-Aldrich®
Tetrakis(dimethylamido)titan
Formel: C₈H₂₄N₄Ti
Molecular Weight: 224,17 g/mol
Flammpunkt: –30 °C (geschlossener Tiegel)
MDL: MFCD00014861
CAS-Nummer: 3275-24-9
EINECS: 221-904-3
UN: 3129
ADR: 4.3,II

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Light Yellow to Dark Yellow and Light
Appearance (Form) Liquid
Proton NMR Spectrum Conforms to Structure
Chloride by Ion Chromatography < 10 ppm
Purity Meets Requirements 99.999% Based On Trace Metals Analysis
Trace Metal Analysis < 20.0 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR