Tetrakis(dimethylamino)zirkonium(IV), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
669016-25G
SIAL669016-25GEA
1720
EUR
SIAL669016-25G
Tetrakis(dimethylamino)zirkonium(IV), Sigma-Aldrich®
Tetrakis(dimethylamino)zirkonium(IV)
Formel:
C₈H₂₄N₄Zr |
MDL:
MFCD00239502 CAS-Nummer: 19756-04-8 UN: 3396 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...