Methyl-α-D-glucopyranosid, Millipore®

Lieferant: Merck

Synonyme: α-D-Methylglucoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol

66940-100G
SIAL66940-100GEA 40.6 EUR
SIAL66940-100G
Methyl-α-D-glucopyranosid, Millipore®
Methyl-α-D-glucopyranosid
Formel: C₇H₁₄O₆
Molecular Weight: 194,18 g/mol
MDL: MFCD00064086
CAS-Nummer: 97-30-3
EINECS: 202-571-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR