2,3'-Dibrom-4'-fluoracetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Bromo-1-(3-bromo-4-fluorophenyl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

669458-1G
SIAL669458-1GEA 85.2 EUR
SIAL669458-1G
2,3'-Dibrom-4'-fluoracetophenon, Sigma-Aldrich®
2,3'-Dibrom-4'-fluoracetophenon
Formel: C₈H₅Br₂FO
Molecular Weight: 295,93 g/mol
MDL: MFCD07368759
CAS-Nummer: 435273-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR