ω-Brom-3-fluoracetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Brommethyl-m-fluorphenylketon, 2-Brom-3'-fluoracetophenon, 2-Brom-1-(3-fluorphenyl)ethan-1-on, α-Brom-3-fluoracetophenon, 3-Fluorphenacylbromid

M-Clarity™ Qualitätsstufe = MQ100

669482-1G
SIAL669482-1GEA 92.1 EUR
SIAL669482-1G
ω-Brom-3-fluoracetophenon, Sigma-Aldrich®
ω-Brom-3-fluoracetophenon
Formel: C₈H₆BrFO
Molecular Weight: 217.04 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00109756
CAS-Nummer: 53631-18-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR