Methyl-6-hydroxy-5-nitronicotinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 6-hydroxy-5-nitronicotinate, Methyl 6-hydroxy-5-nitropyridine-3-carboxylate

669784-5G
SIAL669784-5GEA 527 EUR
SIAL669784-5G
Methyl-6-hydroxy-5-nitronicotinat, Sigma-Aldrich®
Methyl-6-hydroxy-5-nitronicotinat
Formel: C₇H₆N₂O₅
Molecular Weight: 198,14 g/mol
MDL: MFCD10699641
CAS-Nummer: 222970-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR