5-Brom-1-pentanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromopentan-1-ol

M-Clarity™ Qualitätsstufe = MQ100

670855-5G
SIAL670855-5GEA 95.3 EUR
SIAL670855-5G
5-Brom-1-pentanol, Sigma-Aldrich®
5-Bromo-1-pentanol
Formel: C₅H₁₁BrO
Molecular Weight: 167.05 g/mol
MDL: MFCD00061101
CAS-Nummer: 34626-51-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR