(1S,2S)-(-)-2-Amino-1,2-diphenylethanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

671223-500MG
SIAL671223-500MGEA 207 EUR
SIAL671223-500MG
(1S,2S)-(-)-2-Amino-1,2-diphenylethanol, Sigma-Aldrich®
(1S,2S)-(-)-2-Amino-1,2-diphenylethanol
Formel: C₁₄H₁₅NO
Molecular Weight: 213,28 g/mol
CAS-Nummer: 23190-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR