Propionohydrazid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Propionhydrazid, Propionsäurehydrazid
M-Clarity™ Qualitätsstufe = MQ100
671460-1G
SIAL671460-1GEA
37.8
EUR
SIAL671460-1G
Propionohydrazid, Sigma-Aldrich®
Propionohydrazid
Formel:
C₃H₈N₂O Molecular Weight: 88.11 g/mol |
MDL:
MFCD01333200 CAS-Nummer: 5818-15-5 EINECS: 227-390-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...