4'-Chlor-3'-methylacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(4-Chloro-3-methylphenyl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

673153-10G
SIAL673153-10GEA 136 EUR
SIAL673153-10G
4'-Chlor-3'-methylacetophenon, Sigma-Aldrich®
4'-Chlor-3'-methylacetophenon
Formel: C₉H₉ClO
Molecular Weight: 168,62 g/mol
MDL: MFCD00045172
CAS-Nummer: 37074-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR