2,6-Dichlor-4-nitropyridin-N-oxid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

673161-5G
SIAL673161-5GEA 93.7 EUR
SIAL673161-5G
2,6-Dichlor-4-nitropyridin-N-oxid, Sigma-Aldrich®
2,6-Dichlor-4-nitropyridin-N-oxid
Formel: C₅H₂Cl₂N₂O₃
Molecular Weight: 208,99 g/mol
MDL: MFCD00955611
CAS-Nummer: 2587-01-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR