1,5-Naphthyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

673218-1G
SIAL673218-1GEA 136 EUR
SIAL673218-1G
1,5-Naphthyridin, Sigma-Aldrich®
1,5-Naphthyridine
Formel: C₈H₆N₂
Molecular Weight: 130,15 g/mol
MDL: MFCD00039727
CAS-Nummer: 254-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR