4-Nitrophenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

673854-1G 673854-5G
SIAL673854-1GEA 78.4 EUR
SIAL673854-1G SIAL673854-5G
4-Nitrophenylboronsäure, Sigma-Aldrich®
4-Nitrophenylboronsäure
Formel: C₆H₆BNO₄
MDL: MFCD00161360
CAS-Nummer: 24067-17-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR