2-Nitrophenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

673862-5G
SIAL673862-5GEA 0 EUR
SIAL673862-5G
2-Nitrophenylboronsäure, Sigma-Aldrich®
2-Nitrophenylboronsäure
Formel: C₆H₆BNO₄
MDL: MFCD00161358
CAS-Nummer: 5570-19-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR