(±)-2,3-Difluormandelsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-2,3-Difluorphenylglykolsäure, (±)-2-(2,3-Difluorphenyl)-2-hydroxyethansäure, (±)-2-(2,3-Difluorphenyl)-2-hydroxyessigsäure

M-Clarity™ Qualitätsstufe = MQ100

674001-1G
SIAL674001-1GEA 85.1 EUR
SIAL674001-1G
(±)-2,3-Difluormandelsäure, Sigma-Aldrich®
(±)-2,3-Difluormandelsäure
Formel: C₈H₆F₂O₃
Molecular Weight: 188,13 g/mol
MDL: MFCD00061295
CAS-Nummer: 207974-19-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR