Poly(methacrylic acid, sodium salt) solution, M.W. 4000 - 6000 40% (w/w) in Wasser, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

674044-1L
SIAL674044-1LEA 112 EUR
SIAL674044-1L
Poly(methacrylic acid, sodium salt) solution, M.W. 4000 - 6000 40% (w/w) in Wasser, Sigma-Aldrich®
Poly(methacrylic acid, sodium salt) solution
Formel: C₄H₅NaO₂
Dichte: 1,251 g/ml (25 °C)
MDL: MFCD00156535
CAS-Nummer: 54193-36-1

Bestellen

Spezifikation Testergebnisse

Infrared spectrum Conforms to structure
Solid content 38.0 - 42.0%
Concentration 40 wt.% in H₂O
Refractive index n20/D 1.430


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR