4-Chlor-3-nitropyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

674079-5G
SIAL674079-5GEA 62.8 EUR
SIAL674079-5G
4-Chlor-3-nitropyridin, Sigma-Aldrich®
4-Chloro-3-nitropyridine
Formel: C₅H₃ClN₂O₂
MDL: MFCD02683025
CAS-Nummer: 13091-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR