11-Mercaptoundecansäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

674427-500MG
SIAL674427-500MGEA 494 EUR
SIAL674427-500MG
11-Mercaptoundecansäure, Sigma-Aldrich®
11-Mercaptoundecanoic acid
Formel: C₁₁H₂₂O₂S
MDL: MFCD00022096
CAS-Nummer: 71310-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR