Benzo[b]thiophen-2-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Benzo[b]thiophene-2-carbaldehyde

M-Clarity™ Qualitätsstufe = MQ100

674540-5G
SIAL674540-5GEA 26.3 EUR
SIAL674540-5G
Benzo[b]thiophen-2-carbaldehyd, Sigma-Aldrich®
Benzo[b]thiophen-2-carbaldehyd
Formel: C₉H₆OS
Molecular Weight: 162,21 g/mol
MDL: MFCD01075041
CAS-Nummer: 3541-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR