(R)-(+)-1-Boc-3-methylpiperazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (3R)-3-Methylpiperazine, N1-BOC protected, (R)-1-Boc-3-Methylpiperazine, (R)-1-Boc-3-methyl-piperazine

M-Clarity™ Qualitätsstufe = MQ100

674761-1G
SIAL674761-1GEA 78.5 EUR
SIAL674761-1G
(R)-(+)-1-Boc-3-methylpiperazin, Sigma-Aldrich®
(R)-(+)-1-Boc-3-methylpiperazin
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,28 g/mol
MDL: MFCD02683205
CAS-Nummer: 163765-44-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR