4-Methyl-2-(methylthio)pyrimidin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Methyl-2(methylsulphanyl)pyrimidine
M-Clarity™ Qualitätsstufe = MQ100
674885-10G
SIAL674885-10GEA
83.3
EUR
SIAL674885-10G
4-Methyl-2-(methylthio)pyrimidin, Sigma-Aldrich®
4-Methyl-2-(methylthio)pyrimidine
Formel:
C₆H₈N₂S Molecular Weight: 140,21 g/mol |
MDL:
MFCD00023242 CAS-Nummer: 14001-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...