7-Chlorisatin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 7-Chlorindolin-2,3-dion, 7-Chlor-2,3-dioxoindolin, 7-Chlor-1H-indol-2,3-dion, 7-Chlor-2,3-indolindion

675040-1G
SIAL675040-1GEA 76.4 EUR
SIAL675040-1G
7-Chlorisatin, Sigma-Aldrich®
7-Chlorisatin
Formel: C₈H₄ClNO₂
MDL: MFCD00022796
CAS-Nummer: 7477-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR