Isochinolin-5-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Isoquinoline-5-carboxaldehyde

675180-1G
SIAL675180-1GEA 75.3 EUR
SIAL675180-1G
Isochinolin-5-carbaldehyd, Sigma-Aldrich®
Isochinolin-5-carbaldehyd
Formel: C₁₀H₇NO
Molecular Weight: 157,17 g/mol
MDL: MFCD03412483
CAS-Nummer: 80278-67-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR