4,6-Dimethoxy-o-toluylsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4-Dimethoxy-6-methylbenzoesäure

M-Clarity™ Qualitätsstufe = MQ100

675253-1G
SIAL675253-1GEA 133 EUR
SIAL675253-1G
4,6-Dimethoxy-o-toluylsäure, Sigma-Aldrich®
4,6-Dimethoxy-o-toluylsäure
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
MDL: MFCD00229046
CAS-Nummer: 3686-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR