3,5-Dimethyl-4-hydroxybenzonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Hydroxy-3,5-dimethylbenzonitrile

M-Clarity™ Qualitätsstufe = MQ100

675385-1G
SIAL675385-1GEA 44.9 EUR
SIAL675385-1G
3,5-Dimethyl-4-hydroxybenzonitril, Sigma-Aldrich®
3,5-Dimethyl-4-hydroxybenzonitril
Formel: C₉H₉NO
Molecular Weight: 147,18 g/mol
MDL: MFCD00221716
CAS-Nummer: 4198-90-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR