1-Methyl-2-(tributylstannyl)pyrrol, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

675679-1G
SIAL675679-1GEA 109 EUR
SIAL675679-1G
1-Methyl-2-(tributylstannyl)pyrrol, Sigma-Aldrich®
1-Methyl-2-(tributylstannyl)pyrrol
Formel: C₁₇H₃₃NSn
Molecular Weight: 370,15 g/mol
MDL: MFCD01319022
CAS-Nummer: 118486-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR