(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridine, CTH-(R)-P-Phos

M-Clarity™ Qualitätsstufe = MQ100

675806-100MG
SIAL675806-100MGEA 108 EUR
SIAL675806-100MG
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridin, Sigma-Aldrich®
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(diphenylphosphino)-3,3'-bipyridin
Formel: C₃₈H₃₄N₂O₄P₂
Molecular Weight: 644.64 g/mol
MDL: MFCD04038734
CAS-Nummer: 221012-82-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR