3,5-Dibromanisol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,3-Dibromo-5-methoxybenzene

M-Clarity™ Qualitätsstufe = MQ100

675830-1G
SIAL675830-1GEA 27 EUR
SIAL675830-1G
3,5-Dibromanisol, Sigma-Aldrich®
3,5-Dibromanisol
Formel: C₇H₆Br₂O
Molecular Weight: 265,93 g/mol
MDL: MFCD02258848
CAS-Nummer: 74137-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR