3-Brom-5-fluorphenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Brom-3-fluor-5-hydroxybenzol

M-Clarity™ Qualitätsstufe = MQ100

675857-5G
SIAL675857-5GEA 68.5 EUR
SIAL675857-5G
3-Brom-5-fluorphenol, Sigma-Aldrich®
3-Brom-5-fluorphenol
Formel: C₆H₄BrFO
Molecular Weight: 191 g/mol
MDL: MFCD07783710
CAS-Nummer: 433939-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR