2-Fluor-4-nitroanilin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
675881-5G
SIAL675881-5GEA
52.7
EUR
SIAL675881-5G
2-Fluor-4-nitroanilin, Sigma-Aldrich®
2-Fluor-4-nitroanilin
Formel:
C₆H₅FN₂O₂ Molecular Weight: 156,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00034560 CAS-Nummer: 369-35-7 EINECS: 206-719-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...