4-(Methylsulfonyl)phenylboronsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
675903-1G
SIAL675903-1GEA
37.4
EUR
SIAL675903-1G
4-(Methylsulfonyl)phenylboronsäure, Sigma-Aldrich®
4-(Methylsulfonyl)phenylboronsäure
Formel:
C₇H₉BO₄S |
MDL:
MFCD01630820 CAS-Nummer: 149104-88-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...