1-Boc-2-indolboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(tert-Butoxycarbonyl)-1H-indole-2-boronic acid, 1H-Indole-2-boronic acid, N-BOC protected, N-Boc-indole-2-boronic Acid, 1-(tert-Butoxycarbonyl)indole-2-boronic acid, N-Boc-indole-2-boronic acid

M-Clarity™ Qualitätsstufe = MQ100

675911-1G 675911-5G
SIAL675911-1GEA 58.2 EUR
SIAL675911-1G SIAL675911-5G
1-Boc-2-indolboronsäure, Sigma-Aldrich®
1-Boc-2-indolboronsäure
Formel: C₁₃H₁₆BNO₄
Molecular Weight: 261.08 g/mol
MDL: MFCD02093045
CAS-Nummer: 213318-44-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR